missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Methioninamide hydrochloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 426710050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
L-Methioninamide hydrochloride | |
97.5 to 102.5% (Argentometry) | |
C5H13ClN2OS | |
MFCD00039084 | |
l-methioninamide hydrochloride, h-met-nh2 hcl, h-met-nh2.hcl, s-2-amino-4-methylthio butanamide hydrochloride, 2s-2-amino-4-methylthiobutanamide, chloride, h-met-nh hcl, h-met-nh2??hcl, methioninamide hydrochloride, h-l-met-nh2 hydrochloride, methionine amide hydrochloride | |
Cl.CSCCC(N)C(N)=O | |
184.68 | |
184.69 |
16120-92-6 | |
Glass bottle | |
CH3SCH2CH2CH(CNH2)CONH2·HCl | |
5g | |
PWCBMJHINDTXGV-UHFFFAOYNA-N | |
(2S)-2-amino-4-methylsulfanylbutanamide;hydrochloride | |
18531008 | |
98% |
Chemical Identifiers
16120-92-6 | |
184.68 | |
PWCBMJHINDTXGV-UHFFFAOYNA-N | |
18531008 | |
Cl.CSCCC(N)C(N)=O |
C5H13ClN2OS | |
MFCD00039084 | |
l-methioninamide hydrochloride, h-met-nh2 hcl, h-met-nh2.hcl, s-2-amino-4-methylthio butanamide hydrochloride, 2s-2-amino-4-methylthiobutanamide, chloride, h-met-nh hcl, h-met-nh2??hcl, methioninamide hydrochloride, h-l-met-nh2 hydrochloride, methionine amide hydrochloride | |
(2S)-2-amino-4-methylsulfanylbutanamide;hydrochloride |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only