missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Menthone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1367918

 View more versions of this product

Catalog No. AAA1367918



Description

Description

L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

Solubility
Soluble in ether, alcohol, benzene, acetone.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

L-Menthone
0.893
72°C (162°F)
C10H18O
MFCD00001634,MFCD00136033
3648743
--menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
NFLGAXVYCFJBMK-BDAKNGLRSA-N
(2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
26447
154.25
Liquid
14073-97-3
208°C to 210°C
97%
1.45
50 g
14,5838
Soluble in ether,alcohol,benzene,acetone.
CC(C)[C@@H]1CC[C@@H](C)CC1=O
154.25
CHEBI:15410
97%

Chemical Identifiers

14073-97-3
154.25
NFLGAXVYCFJBMK-BDAKNGLRSA-N
26447
(2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
C10H18O
MFCD00001634,MFCD00136033
--menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
CHEBI:15410
CC(C)[C@@H]1CC[C@@H](C)CC1=O
Safety and Handling

Safety and Handling

GHS H Statement
H319-H227

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 237-926-1

RTECSNumber : OS9542200

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only