missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(+)-Lysine Monohydrochloride, TCI America™

Manufacturer:  TCI America L0071500G

 View more versions of this product

Catalog No. L0071500G



Specifications

Specifications

L-(+)-Lysine Monohydrochloride
500g
C6H15ClN2O2
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
C(CCN)CC(C(=O)O)N.Cl
182.648
CHEBI:53633
Crystal-Powder at 20°C
White
657-27-2
MFCD00064564
BVHLGVCQOALMSV-JEDNCBNOSA-N
(2S)-2,6-diaminohexanoic acid;hydrochloride
69568
182.65
≥98.0% (T)

Chemical Identifiers

657-27-2
182.648
BVHLGVCQOALMSV-JEDNCBNOSA-N
69568
(2S)-2,6-diaminohexanoic acid;hydrochloride
C6H15ClN2O2
MFCD00064564
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
CHEBI:53633
C(CCN)CC(C(=O)O)N.Cl
Safety and Handling

Safety and Handling

RTECSNumber : OL5650000

TSCA : Yes

Documents