missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Lysine Methyl Ester Dihydrochloride, TCI America™

Manufacturer:  TCI America L02025G

 View more versions of this product

Catalog No. L02025G



Specifications

Specifications

L-Lysine Methyl Ester Dihydrochloride
197°C
26348-70-9
MFCD00039067
SXZCBVCQHOJXDR-ILKKLZGPSA-N
methyl (2S)-2,6-diaminohexanoate;dihydrochloride
117778
Crystal-Powder at 20°C
White
5g
C7H18Cl2N2O2
l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
COC(=O)C(CCCCN)N.Cl.Cl
233.133
233.13
≥98.0% (N)

Chemical Identifiers

26348-70-9
233.133
SXZCBVCQHOJXDR-ILKKLZGPSA-N
117778
COC(=O)C(CCCCN)N.Cl.Cl
C7H18Cl2N2O2
MFCD00039067
l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
methyl (2S)-2,6-diaminohexanoate;dihydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents