missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

L-Lysine Hydrochloride (White Powder), Fisher BioReagents
Manufacturer: Fisher BioReagents BP386100
Description
This amino acid is suitable for use in tissue culture systems requiring additives.Specifications
L-Lysine Hydrochloride | |
657-27-2 | |
19.12 to 19.51% | |
98.5 to 100.5 % | |
C6H15ClN2O2 | |
15, 5697 | |
Glass Bottle | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
C(CCN)CC(C(=O)O)N.Cl | |
20.7 to 21.5° (+ or -) | |
69568 | |
182.65 | |
Solid | |
Lysine Hydrochloride | |
263°C |
0.02% max. | |
1ppm max. | |
10ppm max. | |
Pass Test | |
0.4% max. | |
Chromatographically not detectable | |
darvyl, enisyl, h-lys-oh.hcl, l-lysine hydrochloride, l-lysine monohydrochloride, l-lysine, monohydrochloride, lyamine, lysine hydrochloride, lysion, s-2,6-diaminohexanoic acid hydrochloride | |
0.1% max. | |
(2S)-2,6-diaminohexanoic acid;hydrochloride | |
182.648 | |
CHEBI:53633 | |
98.0% min. | |
98.5 to 100.5% | |
White | |
100g |
Chemical Identifiers
657-27-2 | |
182.648 | |
darvyl, enisyl, h-lys-oh.hcl, l-lysine hydrochloride, l-lysine monohydrochloride, l-lysine, monohydrochloride, lyamine, lysine hydrochloride, lysion, s-2,6-diaminohexanoic acid hydrochloride | |
CHEBI:53633 | |
C(CCN)CC(C(=O)O)N.Cl |
C6H15ClN2O2 | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
69568 | |
(2S)-2,6-diaminohexanoic acid;hydrochloride |
Safety and Handling

Recommended Storage : RT