Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Lysine dihydrochloride, 99%

$124.16 - $124.16

Chemical Identifiers

Chemical Name or Material L-Lysine dihydrochloride
CAS 657-26-1
Molecular Formula C6H14N2O2·2HCl
Formula Weight 219.11
MDL Number MFCD00066389
Beilstein 4539715
Merck Index 14,5636
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1612722

  SDS
alfa aesar™
A1612722
100g Each for $124.16
Description & Specifications

Specifications

Formula Weight 219.11
TSCA TSCA
Beilstein 4539715
Molecular Formula C6H14N2O2·2HCl
Melting Point 202° to 206°C
CAS 657-26-1
Chemical Name or Material L-Lysine dihydrochloride
MDL Number MFCD00066389
Quantity 100g
Merck Index 14,5636
Assay 99%

Applications
Synthesis of hyperbranched polylysine using the N-hydroxysuccinimide ester of L-lysine dihydrochloride as AB 2 building block. The histidine and lysine standards were prepared from L-histidine monohydro- chloride and L-lysine dihydrochloride respectively. L-Homocitrulline-The copper complex was prepared from L-lysine dihydrochloride in 80 to 86 per cent yields.

Solubility
Soluble in water (100 mg/ml).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.