missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Lysine acetate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451630250
Specifications
L-Lysine acetate | |
97% | |
l-lysine acetate, l-lysine acetate salt, l-lysine monoacetate, l-lysine, acetate, lysine acetate, lysine acetate salt, lysine acetate usp, lysine, monoacetate, ttl6g7liwz, unii-ttl6g7liwz | |
CC(=O)O.C(CCN)CC(C(=O)O)N | |
206.242 | |
206.24 | |
25g |
57282-49-2 | |
C8H18N2O4 | |
RRNJROHIFSLGRA-JEDNCBNOSA-N | |
acetic acid;(2S)-2,6-diaminohexanoic acid | |
104152 | |
97% |
Chemical Identifiers
57282-49-2 | |
206.242 | |
l-lysine acetate, l-lysine acetate salt, l-lysine monoacetate, l-lysine, acetate, lysine acetate, lysine acetate salt, lysine acetate usp, lysine, monoacetate, ttl6g7liwz, unii-ttl6g7liwz | |
acetic acid;(2S)-2,6-diaminohexanoic acid |
C8H18N2O4 | |
RRNJROHIFSLGRA-JEDNCBNOSA-N | |
104152 | |
CC(=O)O.C(CCN)CC(C(=O)O)N |