missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Lysinamide dihydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H65741MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Lysinamide dihydrochloride | |
C6H17Cl2N3O | |
s-2,6-diaminohexanamide dihydrochloride, h-lys-nh2 2hcl, l-lysinamide dihydrochloride, h-lys-nh 2 hcl, lysine amide dihydrochloride, c6h15n3o.2hcl, h-lys-nh2 . 2 hcl | |
Cl.Cl.NCCCC[C@H](N)C(N)=O | |
218.12 | |
218.12 | |
250 mg |
51127-08-3 | |
MFCD00058287 | |
AIYVJLPYZQDCKV-XRIGFGBMSA-N | |
(2S)-2,6-diaminohexanamide dihydrochloride | |
20760365 | |
95% | |
Solid |
Chemical Identifiers
51127-08-3 | |
218.12 | |
AIYVJLPYZQDCKV-XRIGFGBMSA-N | |
20760365 | |
Cl.Cl.NCCCC[C@H](N)C(N)=O |
C6H17Cl2N3O | |
MFCD00058287 | |
s-2,6-diaminohexanamide dihydrochloride, h-lys-nh2 2hcl, l-lysinamide dihydrochloride, h-lys-nh 2 hcl, lysine amide dihydrochloride, c6h15n3o.2hcl, h-lys-nh2 . 2 hcl | |
(2S)-2,6-diaminohexanamide dihydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only