missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Leucyl-L-tyrosine, TCI America™

Manufacturer:  TCI America L0040100MG

 View more versions of this product

Catalog No. L0040100MG



Specifications

Specifications

L-Leucyl-L-tyrosine
100mg
C15H22N2O4
l-leucyl-l-tyrosine, s-2-s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, chembl56099, leucyl-tyrosine, 2s-2-2s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2s-2-amino-4-methylpentanoyl amino-3-4-hydroxyphenyl propanoic acid, n-l-leucyl-l-tyrosine, tyrosine, leucyl, h-leu-tyr-oh, l-leu-l-tyr
CC(C)CC(C(=O)NC(CC1=CC=C(C=C1)O)C(=O)O)N
294.351
CHEBI:73591
Crystal-Powder at 20°C
White
968-21-8
MFCD00020195
LHSGPCFBGJHPCY-STQMWFEESA-N
(2S)-2-[[(2S)-2-amino-4-methylpentanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
70410
294.35
≥98.0% (T)

Chemical Identifiers

968-21-8
294.351
LHSGPCFBGJHPCY-STQMWFEESA-N
70410
(2S)-2-[[(2S)-2-amino-4-methylpentanoyl]amino]-3-(4-hydroxyphenyl)propanoic acid
C15H22N2O4
MFCD00020195
l-leucyl-l-tyrosine, s-2-s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, chembl56099, leucyl-tyrosine, 2s-2-2s-2-amino-4-methylpentanamido-3-4-hydroxyphenyl propanoic acid, 2s-2-2s-2-amino-4-methylpentanoyl amino-3-4-hydroxyphenyl propanoic acid, n-l-leucyl-l-tyrosine, tyrosine, leucyl, h-leu-tyr-oh, l-leu-l-tyr
CHEBI:73591
CC(C)CC(C(=O)NC(CC1=CC=C(C=C1)O)C(=O)O)N
Safety and Handling

Safety and Handling

TSCA : No

Documents