missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-(+)-Leucinol 97.0+%, TCI America™
Supplier: TCI America L013725ML
Specifications
L-(+)-Leucinol | |
Yellow | |
C6H15NO | |
25 mL | |
VPSSPAXIFBTOHY-LURJTMIESA-N | |
(2S)-2-amino-4-methylpentan-1-ol | |
111307 | |
≥97.0% (GC,T) |
7533-40-6 | |
210°C | |
MFCD00063676 | |
l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol | |
CC(C)C[C@H](N)CO | |
117.19 | |
117.19 | |
Liquid |
Chemical Identifiers
7533-40-6 | |
117.19 | |
VPSSPAXIFBTOHY-LURJTMIESA-N | |
111307 | |
CC(C)C[C@H](N)CO |
C6H15NO | |
MFCD00063676 | |
l-leucinol, s-+-leucinol, h-leucinol, 2s-2-amino-4-methylpentan-1-ol, l-+-leucinol, l +-leucinol, s-2-amino-4-methylpentan-1-ol, leucinol, s-leucinol, 2s-2-amino-4-methyl-1-pentanol | |
(2S)-2-amino-4-methylpentan-1-ol |
Safety and Handling
TSCA : No