missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Leucine tert-butyl ester hydrochloride, 98%

$69.24 - $240.57

Chemical Identifiers

CAS 2749-11-3
Molecular Formula C3H10NO
Molecular Weight (g/mol) 76.12
MDL Number MFCD00038906
InChI Key BKMMTJMQCTUHRP-VKHMYHEASA-O
Synonym l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
PubChem CID 80307
ChEBI CHEBI:78502
IUPAC Name (2S)-2-aminopropan-1-ol
SMILES CC(CO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5998003 View Documents Alfa Aesar™
H5998003
1g Each for $69.24
 
AAH5998006 View Documents Alfa Aesar™
H5998006
5g Each for $240.57
 
Specifications

Chemical Identifiers

2749-11-3
76.12
BKMMTJMQCTUHRP-VKHMYHEASA-O
80307
(2S)-2-aminopropan-1-ol
C3H10NO
MFCD00038906
l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
CHEBI:78502
CC(CO)N

Specifications

2748-02-9
MFCD00038906
h-leu-otbu.hcl, l-leucine tert-butyl ester hydrochloride, h-leu-otbu hcl, l-leucine t-butyl ether hcl salt, l-leucine t-butyl ester hydrochloride, l-leucine tert-butyl ester hcl, tert-butyl l-leucinate hydrochloride, s-tert-butyl 2-amino-4-methylpentanoate hydrochloride, tert-butyl 2s-2-amino-4-methylpentanoate hydrochloride, h-leu-obut hcl
CC(C)CC(C(=O)OC(C)(C)C)N.Cl
223.74
223.74
1g
L-Leucine tert-butyl ester hydrochloride
C10H22ClNO2
3697141
RFUWRXIYTQGFGA-QRPNPIFTSA-N
tert-butyl (2S)-2-amino-4-methylpentanoate;hydrochloride
12451443
98%
+12° (c=2 in DMF)
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C