missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Leucine methyl ester hydrochloride, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals A1489736
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
L-Leucine methyl ester hydrochloride | |
C7H16ClNO2 | |
3595133 | |
DODCBMODXGJOKD-RGMNGODLSA-N | |
methyl (2S)-2-amino-4-methylpentanoate;hydrochloride | |
2723695 | |
99% | |
+13° (c=2 in Water) |
7517-19-3 | |
MFCD00012494 | |
l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride | |
CC(C)CC(C(=O)OC)N.Cl | |
181.66 | |
181.66 | |
500g | |
Hygroscopic |
Safety and Handling
EINECSNumber : 231-375-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO â Research Use Only