Please login to your online account to display your discounted pricing

L-Leucine ethyleester hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-Leucine ethyl ester hydrochloride
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H17NO2·ClH
Formula Weight 195.69
Synonym Leu-OEt.HCl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC442710050

  SDS
thermo fisher scientific
AC442710050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442710250

  SDS
thermo fisher scientific
AC442710250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 195.69
Quantity 5g
Color White
Melting Point 130° to 140°C
Chemical Name or Material L-Leucine ethyl ester hydrochloride
Assay Percent Range 99%
Molecular Formula C8H17NO2·ClH
CAS 2743-40-0
Infrared Spectrum Authentic
Synonym Leu-OEt.HCl
Packaging Glass bottle
Specific Rotation +17.0° to +19.0° (20°C, 589nm) (c=2, ethanol)