Please login to your online account to display your discounted pricing

L-Leucine ethyleester hydrochloride, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC442710250

 View more versions of this product

Catalog No. AC442710250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 195.69
Quantity 25g
Color White
Melting Point 130° to 140°C
Chemical Name or Material L-Leucine ethyl ester hydrochloride
Synonym Leu-OEt.HCl
Assay Percent Range 99%
Molecular Formula C8H17NO2·ClH
CAS 2743-40-0
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +17.0° to +19.0° (20°C, 589nm) (c=2, ethanol)

Specifications

Chemical Name or Material L-Leucine ethyl ester hydrochloride
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H17NO2·ClH
Formula Weight 195.69
Synonym Leu-OEt.HCl