missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Leucine Ethyl Ester Hydrochloride, TCI America™

Manufacturer:  TCI America L0123250G

 View more versions of this product

Catalog No. L0123250G



Specifications

Specifications

L-Leucine Ethyl Ester Hydrochloride
136°C
2743-40-0
MFCD00034879
NOUDPBCEONUCOV-FJXQXJEOSA-N
ethyl (2S)-2-amino-4-methylpentanoate;hydrochloride
11658447
Crystal-Powder at 20°C
White
250g
C8H18ClNO2
l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl
CCOC(=O)C(CC(C)C)N.Cl
195.687
195.69
≥99.0% (T)

Chemical Identifiers

2743-40-0
195.687
NOUDPBCEONUCOV-FJXQXJEOSA-N
11658447
CCOC(=O)C(CC(C)C)N.Cl
C8H18ClNO2
MFCD00034879
l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl
ethyl (2S)-2-amino-4-methylpentanoate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents