missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Leucine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™
$68.02 - $197.43
Chemical Identifiers
CAS | 1738-77-8 |
---|---|
Molecular Formula | C20H27NO5S |
Molecular Weight (g/mol) | 393.50 |
MDL Number | MFCD00066117 |
InChI Key | QTQGHKVYLQBJLO-YDALLXLXSA-N |
Synonym | l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh |
PubChem CID | 6451312 |
IUPAC Name | 4-methylbenzene-1-sulfonic acid; benzyl (2S)-2-amino-4-methylpentanoate |
SMILES | CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
1738-77-8 | |
393.50 | |
QTQGHKVYLQBJLO-YDALLXLXSA-N | |
6451312 | |
CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC1=CC=CC=C1 |
C20H27NO5S | |
MFCD00066117 | |
l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh | |
4-methylbenzene-1-sulfonic acid; benzyl (2S)-2-amino-4-methylpentanoate |
Specifications
1738-77-8 | |
White | |
MFCD00066117 | |
l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh | |
CC1=CC=C(C=C1)S(O)(=O)=O.CC(C)C[C@H](N)C(=O)OCC1=CC=CC=C1 | |
393.50 | |
393.50 | |
Crystalline Powder |
158°C | |
C20H27NO5S | |
5 g | |
QTQGHKVYLQBJLO-YDALLXLXSA-N | |
4-methylbenzene-1-sulfonic acid; benzyl (2S)-2-amino-4-methylpentanoate | |
6451312 | |
≥98.0% (HPLC,T) | |
L-Leucine Benzyl Ester p-Toluenesulfonate |
Safety and Handling
TSCA : No