missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Leucine benzyl ester p-toluenesulfonate, 98%

$50.89 - $1,016.26

Chemical Identifiers

CAS 1738-77-8
Molecular Formula C20H27NO5S
Molecular Weight (g/mol) 393.498
MDL Number MFCD00066117
InChI Key QTQGHKVYLQBJLO-YDALLXLXSA-N
Synonym l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh
PubChem CID 6451312
IUPAC Name benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.CC(C)CC(C(=O)OCC1=CC=CC=C1)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6247403 View Documents Alfa Aesar™
H6247403
1g Each for $50.89
 
AAH6247406 View Documents Alfa Aesar™
H6247406
5g Each for $226.69
 
AAH6247414 View Documents Alfa Aesar™
H6247414
25g Each for $1,016.26
 
Specifications

Chemical Identifiers

1738-77-8
393.498
QTQGHKVYLQBJLO-YDALLXLXSA-N
6451312
CC1=CC=C(C=C1)S(=O)(=O)O.CC(C)CC(C(=O)OCC1=CC=CC=C1)N
C20H27NO5S
MFCD00066117
l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh
benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid

Specifications

1738-77-8
MFCD00066117
l-leucine benzyl ester p-toluenesulfonate salt, l-leucine benzyl ester p-toluenesulfonate, h-leu-obzl.tosoh, l-leucine benzyl ester tosylate, o-benzyl-l-leucine toluene-p-sulphonate, s-benzyl 2-amino-4-methylpentanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-4-methylpentanoate; para-toluene sulfonate, leu-obzl tosoh, h-leu-obzl.tos, h-leu-obzl tsoh
CC1=CC=C(C=C1)S(=O)(=O)O.CC(C)CC(C(=O)OCC1=CC=CC=C1)N
393.498
393.5
156°C to 160°C
L-Leucine benzyl ester p-toluenesulfonate
C20H27NO5S
3566840
QTQGHKVYLQBJLO-YDALLXLXSA-N
benzyl (2S)-2-amino-4-methylpentanoate;4-methylbenzenesulfonic acid
6451312
98%
1g