missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-leucine, 99.42%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210215801
Specifications
L-Leucine | |
White | |
C6H13NO2 | |
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid | |
CC(C)C[C@H](N)C(O)=O | |
(2S)-2-amino-4-methylpentanoic acid | |
131.18 | |
CHEBI:15603 | |
Small Lustrous Plates or Crystalline Powder |
61-90-5 | |
0.9942 | |
MFCD00002617 | |
ROHFNLRQFUQHCH-MLHKIVSYNA-N | |
1.293g/mL | |
25 g | |
6106 | |
131.18 |
Chemical Identifiers
61-90-5 | |
131.18 | |
ROHFNLRQFUQHCH-MLHKIVSYNA-N | |
6106 | |
CC(C)C[C@H](N)C(O)=O |
C6H13NO2 | |
MFCD00002617 | |
l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid | |
CHEBI:15603 |