Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Leucinamide hydrochloride, 99%

$31.95 - $31.95

Chemical Identifiers

Chemical Name or Material L-Leucinamide hydrochloride
CAS 10466-61-2
Molecular Formula C6H14N2O·HCl
Formula Weight 166.65
MDL Number MFCD00013012
Beilstein 4237021
Synonym (R)-2-Amino-4-methylvaleramide; H-Leu-NH2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2291403

  SDS
alfa aesar™
B2291403
1g Each for $31.95
Description & Specifications

Specifications

Formula Weight 166.65
CAS 10466-61-2
Chemical Name or Material L-Leucinamide hydrochloride
Quantity 1g
Beilstein 4237021
Optical Rotation +9.5° (c=2 in Water)
Synonym (R)-2-Amino-4-methylvaleramide; H-Leu-NH2
Molecular Formula C6H14N2O·HCl
MDL Number MFCD00013012
Assay 99%
Melting Point ≈240°C (dec.)