missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(-)-Lactide, TCI America™

Manufacturer:  TCI America L011525G

 View more versions of this product

Catalog No. L011525G



Specifications

Specifications

L-(-)-Lactide
97°C
4511-42-6
MFCD00070594
JJTUDXZGHPGLLC-IMJSIDKUSA-N
(3S,6S)-3,6-dimethyl-1,4-dioxane-2,5-dione
107983
Crystal-Powder at 20°C
White
25g
C6H8O4
l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1
CC1C(=O)OC(C(=O)O1)C
144.126
144.13
≥98.0% (T)

Chemical Identifiers

4511-42-6
144.126
JJTUDXZGHPGLLC-IMJSIDKUSA-N
107983
CC1C(=O)OC(C(=O)O1)C
C6H8O4
MFCD00070594
l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1
(3S,6S)-3,6-dimethyl-1,4-dioxane-2,5-dione
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5295

TSCA : Yes

Documents