missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Lactide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0903106

 View more versions of this product

Catalog No. AAL0903106



Description

Description

L-Lactide upon polymerization gives poly(lactic acid), which finds many useful properties like biodegradable and nontoxic to the human body and the environment. Its polymer, poly-l-lactide is utilized in the repairing of orbital posterior floor fracture. Further, it is used as biomedical materials for tissue regeneration, matrices for drug delivery systems and alternatives for commercial polymeric materials to reduce the impact on the environment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform, and methanol. Slightly soluble in benzene.

Notes
Moisture sensitive. Store in a cool place. Incompatible with oxidizing agents.
Specifications

Specifications

3.0
95°C to 99°C
255°C
1.4475
MFCD00070594
l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1
JJTUDXZGHPGLLC-UHFFFAOYNA-N
3,6-dimethyl-1,4-dioxane-2,5-dione
107983
≥98%
L-Lactide
4511-42-6
5 g
C6H8O4
82124
Soluble in chloroform,and methanol. Slightly soluble in benzene.
CC1OC(=O)C(C)OC1=O
144.13
144.13

Chemical Identifiers

4511-42-6
144.13
JJTUDXZGHPGLLC-UHFFFAOYNA-N
107983
C6H8O4
MFCD00070594
l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1
CC1OC(=O)C(C)OC1=O
Safety and Handling

Safety and Handling

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-832-0

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only