Please login to your online account to display your discounted pricing

L-(+)-Lactic Acid (Free Acid), Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP26615

 View more versions of this product

Catalog No. BP26615

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 53°C
Boiling Point 119°C
Physical Form Powder/Solid
Formula Weight 90.08
Quantity 5g
Merck Index 15, 5385
Vapor Pressure 0.0813mmHg at 25°C
pH 1.75
Color White
Density 1.20g/cm³
Chemical Name or Material L-(+)-Lactic Acid
Specific Rotation +0.6 to +4.6
Molecular Formula C3H6O3
Infrared Spectrum Conforms to reference
Recommended Storage 0°C, desiccate
Purity 98% min.
Water 1% max.
Name Note Free acid
Assay Percent Range ≥98 %
CAS 79-33-4
ChemAlert Storage Symbol Gray
Synonym (2-Hydroxpropionic acid) Free acid
Identification Pass Test
Packaging Amber Glass

Specifications

Chemical Name or Material L-(+)-Lactic Acid
Name Note Free acid
CAS 79-33-4
Physical Form Powder/Solid
Assay Percent Range ≥98 %
Molecular Formula C3H6O3
Formula Weight 90.08
Synonym (2-Hydroxpropionic acid) Free acid
Accessories
 

0°C, desiccate

SDS & Safety Documents