missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-(+)-Lactic Acid (Free Acid), Fisher BioReagents

$382.83 - $382.83

Chemical Identifiers

CAS 79-33-4
Physical Form Powder/Solid
Assay Percent Range ≥98 %
Molecular Formula C3H6O3
Formula Weight 90.08
Synonym (2-Hydroxpropionic acid) Free acid
Chemical Name or Material L-(+)-Lactic Acid, Free acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BP26615 SDS Fisher Scientific Company
BP26615
Amber Glass 5g Each for $382.83
Case of 9 Each for $3,445.47
Specifications

Description

L-(+)-Lactic Acid is a biochemical that contains varying amounts of intermolecular esterification products.
Description & Specifications

Chemical Identifiers

79-33-4
≥98 %
90.08
L-(+)-Lactic Acid, Free acid
Powder/Solid
C3H6O3
(2-Hydroxpropionic acid) Free acid

Specifications

Powder/Solid
90.08
5g
White
1.75
+0.6 to +4.6
98% min.
Amber Glass
≥98 %
1% max.
L-(+)-Lactic Acid, Free acid
119°C
53°C
15, 5385
1.20g/cm³
0.0813mmHg at 25°C
Conforms to reference
C3H6O3
(2-Hydroxpropionic acid) Free acid
Pass Test
79-33-4

Safety and Handling

Recommended Storage : 0°C, desiccate