missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Isoserine, 98%

$91.61 - $297.64

Chemical Identifiers

CAS 632-13-3
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD03428326
InChI Key BMYNFMYTOJXKLE-REOHCLBHSA-N
Synonym l-isoserine, s-2-hydroxy-b-alanine, s-isoserine, s-3-amino-2-hydroxypropanoic acid, s-3-amino-2-hydroxypropionic acid, 2s-3-amino-2-hydroxypropanoic acid, propanoic acid, 3-amino-2-hydroxy-, 2s, s-isoserine, s-2-hydroxy-, a-alanine, s-2-hydroxy-beta-alanine
PubChem CID 6999797
IUPAC Name (2S)-3-amino-2-hydroxypropanoic acid
SMILES C(C(C(=O)O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60945MD View Documents Alfa Aesar™
H60945MD
250mg Each for $91.61
 
AAH6094503 View Documents Alfa Aesar™
H6094503
1g Each for $297.64
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.
Specifications

Chemical Identifiers

632-13-3
105.093
BMYNFMYTOJXKLE-REOHCLBHSA-N
6999797
C(C(C(=O)O)O)N
C3H7NO3
MFCD03428326
l-isoserine, s-2-hydroxy-b-alanine, s-isoserine, s-3-amino-2-hydroxypropanoic acid, s-3-amino-2-hydroxypropionic acid, 2s-3-amino-2-hydroxypropanoic acid, propanoic acid, 3-amino-2-hydroxy-, 2s, s-isoserine, s-2-hydroxy-, a-alanine, s-2-hydroxy-beta-alanine
(2S)-3-amino-2-hydroxypropanoic acid

Specifications

632-13-3
MFCD03428326
BMYNFMYTOJXKLE-REOHCLBHSA-N
(2S)-3-amino-2-hydroxypropanoic acid
6999797
98%
250mg
C3H7NO3
l-isoserine, s-2-hydroxy-b-alanine, s-isoserine, s-3-amino-2-hydroxypropanoic acid, s-3-amino-2-hydroxypropionic acid, 2s-3-amino-2-hydroxypropanoic acid, propanoic acid, 3-amino-2-hydroxy-, 2s, s-isoserine, s-2-hydroxy-, a-alanine, s-2-hydroxy-beta-alanine
C(C(C(=O)O)O)N
105.093
105.09
197°C to 198°C
L-Isoserine