missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Isoleucinol 97.0+%, TCI America™

$57.61 - $169.53

Chemical Identifiers

CAS 24629-25-2
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00004731
InChI Key VTQHAQXFSHDMHT-NTSWFWBYSA-N
Synonym 1-pentanol, 2-amino-3-methyl-, 2s,3s
PubChem CID 2724027
IUPAC Name (2S,3S)-2-amino-3-methylpentan-1-ol
SMILES CCC(C)C(CO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04621G View Documents Tci America
I04621G
1g Each for $57.61
 
I04625G View Documents Tci America
I04625G
5g Each for $169.53
 
Specifications

Chemical Identifiers

24629-25-2
117.192
VTQHAQXFSHDMHT-NTSWFWBYSA-N
2724027
CCC(C)C(CO)N
C6H15NO
MFCD00004731
1-pentanol, 2-amino-3-methyl-, 2s,3s
(2S,3S)-2-amino-3-methylpentan-1-ol

Specifications

97°C
42°C
24629-25-2
MFCD00004731
VTQHAQXFSHDMHT-NTSWFWBYSA-N
(2S,3S)-2-amino-3-methylpentan-1-ol
2724027
Crystal-Powder at 20°C
L-Isoleucinol
White-Yellow
1g
C6H15NO
1-pentanol, 2-amino-3-methyl-, 2s,3s
CCC(C)C(CO)N
117.192
117.19
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No