Please login to your online account to display your discounted pricing

L(+)-Isoleucinol, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  183940010

 View more versions of this product

Catalog No. AC183940010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Viscous Liquid
Formula Weight 117.19
Quantity 1g
Color White to light yellow
EINECS Number 246-371-4
Specific Rotation Condition + 4.50 (20.00°C c=1.6 C2H5OH)
Chemical Name or Material L(+)-Isoleucinol
Assay Percent Range 97%
Synonym 2-Amino-3-methyl-1-pentanol
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning
Refractive Index 1.4570 to 1.4600 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Specific Rotation +3.5° to +5.5° (20°C, 589nm) (c=1.6, C2H5OH)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Molecular Formula C6H15NO
Linear Formula C2H5CH(CH3)CH(NH2)CH2OH
MDL Number MFCD00004731
CAS 24629-25-2

Specifications

Chemical Name or Material L(+)-Isoleucinol
Name Note 97%
CAS 24629-25-2
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H15NO
Linear Formula C2H5CH(CH3)CH(NH2)CH2OH
Formula Weight 117.19
MDL Number MFCD00004731
Synonym 2-Amino-3-methyl-1-pentanol
Accessories

Warning

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents