missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Isoleucinol, 97%

$66.62 - $277.58

Chemical Identifiers

CAS 24629-25-2
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00004731
InChI Key VTQHAQXFSHDMHT-NTSWFWBYSA-N
Synonym l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, h-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
PubChem CID 2724027
IUPAC Name (2S,3S)-2-amino-3-methylpentan-1-ol
SMILES CCC(C)C(CO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6055603 View Documents Alfa Aesar™
H6055603
1g Each for $66.62
 
AAH6055606 View Documents Alfa Aesar™
H6055606
5g Each for $277.58
 
Specifications

Chemical Identifiers

24629-25-2
117.192
VTQHAQXFSHDMHT-NTSWFWBYSA-N
2724027
CCC(C)C(CO)N
C6H15NO
MFCD00004731
l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, h-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
(2S,3S)-2-amino-3-methylpentan-1-ol

Specifications

24629-25-2
97°C
100°C (212°F)
C2H5CH(CH3)CH(NH2)CH2OH
l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, h-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
CCC(C)C(CO)N
117.192
117.19
L-Isoleucinol
36°C to 39°C
1g
C6H15NO
MFCD00004731
VTQHAQXFSHDMHT-NTSWFWBYSA-N
(2S,3S)-2-amino-3-methylpentan-1-ol
2724027
97%