Please login to your online account to display your discounted pricing

L-Isoleucine, Multi-Compendial, U.S.P., J.T.Baker™


Quantity 100g
Packaging Poly Bottle

Manufacturer: avantor performance  208205

 View more versions of this product

Catalog No. 02003206

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Multi-Compendial/USP
Formula Weight 131.17
Chemical Name or Material L-Isoleucine
CAS 73-32-5
Packaging Poly Bottle
Quantity 100g
Molecular Formula C6H13NO2

Specifications

Chemical Name or Material L-Isoleucine
CAS 73-32-5
Purity Grade Multi-Compendial/USP
Molecular Formula C6H13NO2
Formula Weight 131.17