missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Isoleucine, Multi-Compendial, U.S.P., J.T. Baker™
Supplier: Avantor J.T.Baker 208206
Specifications
L-Isoleucine | |
C6H13NO2 | |
Poly Bottle | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
(2S,3S)-2-amino-3-methylpentanoic acid | |
6306 | |
131.17 | |
1 kg |
73-32-5 | |
MFCD00064222 MFCD00004268 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
CC[C@H](C)[C@H](N)C(O)=O | |
131.18 | |
CHEBI:17191 | |
Multi-Compendial/USP |
Chemical Identifiers
73-32-5 | |
131.18 | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
6306 | |
(2S,3S)-2-amino-3-methylpentanoic acid |
C6H13NO2 | |
MFCD00064222 MFCD00004268 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
CHEBI:17191 | |
CC[C@H](C)[C@H](N)C(O)=O |