missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Isoleucine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6464006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
L-Isoleucine ethyl ester hydrochloride | |
White | |
MFCD09953729 | |
l-isoleucine ethyl ester hydrochloride, h-ile-oet.hcl, ethyl l-isoleucinate hydrochloride, l-isoleucine ethyl ester hcl, 2s,3s-ethyl 2-amino-3-methylpentanoate hydrochloride, isoleucine, ethyl ester, hydrochloride, ethyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, l-isoleucine ethylester hcl, ksc497o6p, l-isoleucineethylesterhydrochloride | |
Cl.CCOC(=O)[C@@H](N)[C@@H](C)CC | |
195.69 | |
195.69 | |
5 g |
56782-52-6 | |
C8H18ClNO2 | |
4710859 | |
QQGRWNMNWONMOO-LEUCUCNGSA-N | |
hydrogen ethyl (2S,3S)-2-amino-3-methylpentanoate chloride | |
6453390 | |
98% | |
Powder |
Chemical Identifiers
56782-52-6 | |
195.69 | |
QQGRWNMNWONMOO-LEUCUCNGSA-N | |
6453390 |
C8H18ClNO2 | |
MFCD09953729 | |
l-isoleucine ethyl ester hydrochloride, h-ile-oet.hcl, ethyl l-isoleucinate hydrochloride, l-isoleucine ethyl ester hcl, 2s,3s-ethyl 2-amino-3-methylpentanoate hydrochloride, isoleucine, ethyl ester, hydrochloride, ethyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, l-isoleucine ethylester hcl, ksc497o6p, l-isoleucineethylesterhydrochloride | |
Cl.CCOC(=O)[C@@H](N)[C@@H](C)CC |
Safety and Handling
EINECSNumber : 260-382-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only