missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Isoleucine, >99%, MP Biomedicals™

$2,026.08 - $2,026.08

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00064222
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym 2-amino-3-methylvaleric acid, 2s,3s-2-amino-3-methylpentanoic acid, 2s,3s-isoleucine, erythro-l-isoleucine, isoleucine, l-+-isoleucine, l-ile, l-isoleucine, s,s-isoleucine, s-isoleucine
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CCC(C)C(C(=O)O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10209291
View Documents
Mp Biomedicals
0210209291
1kg Each for $2,026.08
 
Specifications

Chemical Identifiers

73-32-5
131.175
AGPKZVBTJJNPAG-WHFBIAKZSA-N
6306
(2S,3S)-2-amino-3-methylpentanoic acid
C6H13NO2
MFCD00064222
2-amino-3-methylvaleric acid, 2s,3s-2-amino-3-methylpentanoic acid, 2s,3s-isoleucine, erythro-l-isoleucine, isoleucine, l-+-isoleucine, l-ile, l-isoleucine, s,s-isoleucine, s-isoleucine
CHEBI:17191
CCC(C)C(C(=O)O)N

Specifications

73-32-5
C6H13NO2
2-amino-3-methylvaleric acid, 2s,3s-2-amino-3-methylpentanoic acid, 2s,3s-isoleucine, erythro-l-isoleucine, isoleucine, l-+-isoleucine, l-ile, l-isoleucine, s,s-isoleucine, s-isoleucine
CCC(C)C(C(=O)O)N
131.175
CHEBI:17191
Powder
White
L-Isoleucine
>99%
MFCD00064222
AGPKZVBTJJNPAG-WHFBIAKZSA-N
(2S,3S)-2-amino-3-methylpentanoic acid
6306
131.2
1.003
1kg
Safety and Handling

Safety and Handling

EINECSNumber : 200-798-2

Recommended Storage : Store at Room Temperature (15 to 30°C).