missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Homoserine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1242706
Description
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.
Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Specifications
∽203°C (decomposition) | |
672-15-1 | |
MFCD00063090 | |
14,4739 | |
Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol. | |
C(CO)C(C(=O)O)N | |
119.12 | |
CHEBI:15699 | |
99% | |
−9° (c=2 in Water) |
L-Homoserine | |
C4H9NO3 | |
1721681 | |
l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo | |
UKAUYVFTDYCKQA-VKHMYHEASA-N | |
(2S)-2-amino-4-hydroxybutanoic acid | |
12647 | |
119.12 | |
5 g |
Chemical Identifiers
672-15-1 | |
119.12 | |
UKAUYVFTDYCKQA-VKHMYHEASA-N | |
12647 | |
(2S)-2-amino-4-hydroxybutanoic acid |
C4H9NO3 | |
MFCD00063090 | |
l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo | |
CHEBI:15699 | |
C(CO)C(C(=O)O)N |
Safety and Handling
EINECSNumber : 211-590-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only