Please login to your online account to display your discounted pricing

L-Homoserine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-Homoserine
CAS 672-15-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H9NO3
Formula Weight 119.12
MDL Number MFCD00063090
Synonym L-2-Amino-4-hydroxybutyric acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC204770050

  SDS
thermo fisher scientific
AC204770050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC204770010

  SDS
fisher scientific acros
204770010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 119.12
Quantity 5g
Color White to light yellow
Specific Rotation Condition − 8.50 (22.00°C c=2,H2O)
Beilstein 04, III, 1636
Merck Index 15, 4778
Chemical Name or Material L-Homoserine
Specific Rotation -8° to -9° (22°C, 589nm) (c=2, H2O)
Assay Percent Range 97%
CAS 672-15-1
Molecular Formula C4H9NO3
MDL Number MFCD00063090
Synonym L-2-Amino-4-hydroxybutyric acid
Packaging Glass bottle
Infrared Spectrum Authentic