Please login to your online account to display your discounted pricing

L-β-Homoproline hydrochloride, 98% , ACROS Organics™

Chemical Identifiers

Chemical Name or Material L-β-Homoproline hydrochloride
CAS 53912-85-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H11NO2·ClH
Formula Weight 165.62
MDL Number MFCD07363484
Synonym (S)-2-(2-Pyrrolidinyl)acetic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC426170010

  SDS
thermo fisher scientific
AC426170010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426172500

  SDS
thermo fisher scientific
AC426172500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 165.62
Quantity 1g
Color White to off-white
Chemical Name or Material L-β-Homoproline hydrochloride
Assay Percent Range 98%
Molecular Formula C6H11NO2·ClH
MDL Number MFCD07363484
CAS 53912-85-9
Specific Rotation +31° to +35° (20°C, 589nm) (c=1, H2O)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
NMR Consistent with structure
Packaging Glass bottle
Synonym (S)-2-(2-Pyrrolidinyl)acetic acid hydrochloride
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.