missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Homophenylalanine ethyl ester hydrochloride, 98%

$90.04 - $271.32

Chemical Identifiers

CAS 90891-21-7
Molecular Formula C12H18ClNO2
Molecular Weight (g/mol) 243.731
MDL Number MFCD00190691
InChI Key PTFKZMFFSIYCOV-MERQFXBCSA-N
Synonym ethyl 2s-2-amino-4-phenylbutanoate hydrochloride, ethyl l-homoalaninate hcl, ethyl s-2-amino-4-phenylbutanoate hydrochloride, h-hophe-oet.hcl, h-hph-oet. hcl, h-hph-oet.hcl, homophenylalanine ethyl ester hydrochloride, l-homophenylalanine ethyl ester hydrochloride, pubchem6054, s-ethyl 2-amino-4-phenylbutanoate hydrochloride
PubChem CID 2734606
IUPAC Name ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
SMILES CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6542514 View Documents Alfa Aesar™
H6542514
25g Each for $90.04
 
AAH6542522 View Documents Alfa Aesar™
H6542522
100g Each for $271.32
 
Specifications

Chemical Identifiers

90891-21-7
243.731
PTFKZMFFSIYCOV-MERQFXBCSA-N
2734606
CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
C12H18ClNO2
MFCD00190691
ethyl 2s-2-amino-4-phenylbutanoate hydrochloride, ethyl l-homoalaninate hcl, ethyl s-2-amino-4-phenylbutanoate hydrochloride, h-hophe-oet.hcl, h-hph-oet. hcl, h-hph-oet.hcl, homophenylalanine ethyl ester hydrochloride, l-homophenylalanine ethyl ester hydrochloride, pubchem6054, s-ethyl 2-amino-4-phenylbutanoate hydrochloride
ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride

Specifications

90891-21-7
MFCD00190691
ethyl 2s-2-amino-4-phenylbutanoate hydrochloride, ethyl l-homoalaninate hcl, ethyl s-2-amino-4-phenylbutanoate hydrochloride, h-hophe-oet.hcl, h-hph-oet. hcl, h-hph-oet.hcl, homophenylalanine ethyl ester hydrochloride, l-homophenylalanine ethyl ester hydrochloride, pubchem6054, s-ethyl 2-amino-4-phenylbutanoate hydrochloride
CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
243.731
243.73
98%
159°C to 163°C
L-Homophenylalanine ethyl ester hydrochloride
C12H18ClNO2
6448497
PTFKZMFFSIYCOV-MERQFXBCSA-N
ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
2734606
Powder
White
25g
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold