missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Homophenylalanine ethyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6542522

 View more versions of this product

Catalog No. AAH6542522



Specifications

Specifications

L-Homophenylalanine ethyl ester hydrochloride
C12H18ClNO2
6448497
PTFKZMFFSIYCOV-MERQFXBCSA-N
ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
2734606
Powder
White
100g
90891-21-7
MFCD00190691
ethyl s-2-amino-4-phenylbutanoate hydrochloride, l-homophenylalanine ethyl ester hydrochloride, h-hophe-oet.hcl, ethyl l-homoalaninate hcl, s-ethyl 2-amino-4-phenylbutanoate hydrochloride, ethyl 2s-2-amino-4-phenylbutanoate hydrochloride, pubchem6054, h-hph-oet.hcl, h-hph-oet. hcl, homophenylalanine ethyl ester hydrochloride
CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
243.731
243.73
98%
159°C to 163°C

Chemical Identifiers

90891-21-7
243.731
PTFKZMFFSIYCOV-MERQFXBCSA-N
2734606
CCOC(=O)C(CCC1=CC=CC=C1)N.Cl
C12H18ClNO2
MFCD00190691
ethyl s-2-amino-4-phenylbutanoate hydrochloride, l-homophenylalanine ethyl ester hydrochloride, h-hophe-oet.hcl, ethyl l-homoalaninate hcl, s-ethyl 2-amino-4-phenylbutanoate hydrochloride, ethyl 2s-2-amino-4-phenylbutanoate hydrochloride, pubchem6054, h-hph-oet.hcl, h-hph-oet. hcl, homophenylalanine ethyl ester hydrochloride
ethyl (2S)-2-amino-4-phenylbutanoate;hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents