Please login to your online account to display your discounted pricing

L(+)-Homoarginine hydrochloride, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC169090250

 View more versions of this product

Catalog No. AC169090250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Granular Powder
Assay ≥97.5% (on dried substance)
Formula Weight 224.69
Quantity 25g
Color White
EINECS Number 216-045-6
Packaging Glass bottle
Specific Rotation +17° to +19° (20°C, 589nm) (c=1, 1N HCl)
Water 2% max. (K.F.)
Chemical Name or Material L(+)-Homoarginine hydrochloride
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 98%
Molecular Formula C7H16N4O2·HCl
Linear Formula H2NC(=NH)NH(CH2)4CH(NH2)CO2H·HCl
MDL Number MFCD00012622
CAS 1483-01-8

Specifications

Chemical Name or Material L(+)-Homoarginine hydrochloride
Name Note 99+%
CAS 1483-01-8
Assay ≥97.5% (on dried substance)
Assay Percent Range 98%
Molecular Formula C7H16N4O2·HCl
Linear Formula H2NC(=NH)NH(CH2)4CH(NH2)CO2H·HCl
Formula Weight 224.69
MDL Number MFCD00012622