Learn More
L-Homoarginine hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2738706
Description
An inhibitor of alkaline phosphatase isoenzymes. It is also an inhibitor of cellular transport of arginine by a sodium-independent high affinity y+ transporter. The substitution of homoarginine for arginine or lysine renders proteins resistant to proteolysis by trypsin. Arginine residues are often located at the active centers of proteins and enzymes. Replacing these arginines with homoarginine can elucidate protein function and structural requirements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAn inhibitor of alkaline phosphatase isoenzymes. It is also an inhibitor of cellular transport of arginine by a sodium-independent high affinity y+ transporter. The substitution of homoarginine for arginine or lysine renders proteins resistant to proteolysis by trypsin. Arginine residues are often located at the active centers of proteins and enzymes. Replacing these arginines with homoarginine can elucidate protein function and structural requirements.
Solubility
Soluble in water(50 mg/mL).
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated(2-7°C).
Specifications
213°C to 215°C | |
1483-01-8 | |
MFCD00012622 | |
l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl | |
YMKBVNVCKUYUDM-JEDNCBNOSA-N | |
(2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride | |
2723930 | |
≥98% | |
+18° (c=1 in 1N HCl) |
L-Homoarginine hydrochloride | |
C7H17ClN4O2 | |
4016621 | |
Soluble in water(50mg/mL). | |
C(CCN=C(N)N)CC(C(=O)O)N.Cl | |
224.689 | |
224.69 | |
5 g |
Chemical Identifiers
1483-01-8 | |
224.689 | |
YMKBVNVCKUYUDM-JEDNCBNOSA-N | |
2723930 | |
C(CCN=C(N)N)CC(C(=O)O)N.Cl |
C7H17ClN4O2 | |
MFCD00012622 | |
l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl | |
(2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride |
Safety and Handling
EINECSNumber : 216-045-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only