Please login to your online account to display your discounted pricing

L(-)-Histidinol dihydrochloride, 99%, ACROS Organics™

$208.09 - $208.09

Chemical Identifiers

Chemical Name or Material L(-)-Histidinol dihydrochloride
CAS 1596-64-1
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H11N3O·2HCl
Formula Weight 214.1
MDL Number MFCD00078058
Synonym L-β-Amino-1H-imidazole-4-propanol dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC228830050

  SDS
thermo fisher scientific
AC228830050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC228830010

  SDS
thermo fisher scientific
228830010
Glass bottle 1g Each for $208.09

AC228831000

  SDS
thermo fisher scientific
228831000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 214.1
Physical Form Crystalline Powder
Quantity 5g
Color White to slightly beige
Specific Rotation Condition −3° (24°C c=2,H2O)
Melting Point 194° to 200°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C6H11N3O·2HCl
Chemical Name or Material L(-)-Histidinol dihydrochloride
CAS 1596-64-1
MDL Number MFCD00078058
Synonym L-β-Amino-1H-imidazole-4-propanol dihydrochloride
Packaging Glass bottle
Specific Rotation -2.0° to -3.3° (20°C, 589nm) (c=2, H2O)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.