Please login to your online account to display your discounted pricing

L(-)-Histidinol dihydrochloride, 99%, ACROS Organics™


Quantity 100mg

Manufacturer: thermo fisher scientific  228831000

 View more versions of this product

Catalog No. AC228831000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 214.1
Quantity 100mg
Color White to slightly beige
Specific Rotation Condition −3° (24°C c=2,H2O)
Melting Point 194° to 200°C
Chemical Name or Material L(-)-Histidinol dihydrochloride
Molecular Formula C6H11N3O·2HCl
MDL Number MFCD00078058
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Specific Rotation -2.0° to -3.3° (20°C, 589nm) (c=2, H2O)
Synonym L-β-Amino-1H-imidazole-4-propanol dihydrochloride
Infrared Spectrum Authentic
Assay Percent Range 99%
CAS 1596-64-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material L(-)-Histidinol dihydrochloride
CAS 1596-64-1
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H11N3O·2HCl
Formula Weight 214.1
MDL Number MFCD00078058
Synonym L-β-Amino-1H-imidazole-4-propanol dihydrochloride
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents