Learn More
L-Histidine methyl ester dihydrochloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1238622
Description
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.
Solubility
Soluble in dimethyl sulfoxide, methanol and water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
Specifications
198°C (decomposition) | |
7389-87-9 | |
MFCD00012701 | |
l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl | |
VXXIPPPPKNWFLK-UHFFFAOYNA-N | |
methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl | |
2723645 | |
≥98% | |
+10° (c=2 in water) |
L-Histidine methyl ester dihydrochloride | |
C7H11ClN3O2 | |
3572010 | |
Soluble in dimethyl sulfoxide,methanol and water. | |
[Cl].COC(=O)C(N)CC1=CN=CN1 | |
204.63 | |
242.11 | |
100 g | |
Hygroscopic |
Chemical Identifiers
7389-87-9 | |
204.63 | |
VXXIPPPPKNWFLK-UHFFFAOYNA-N | |
2723645 |
C7H11ClN3O2 | |
MFCD00012701 | |
l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl | |
[Cl].COC(=O)C(N)CC1=CN=CN1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 230-973-9
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only