missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 411735000

 View more versions of this product

Catalog No. AC411735000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L-Histidine hydrochloride monohydrate
200ppm max.
3 to 5 (2.5% soln. at 25°C)
10ppm max.
C6H12ClN3O3
MFCD00151027
25, 513
10, 4617
Plastic Bottle
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
209.63
300ppm max.
98%
+ 9.90 (20.00°C c=11,6 N HCl)
Crystalline Powder or Crystals
98%
5934-29-2
White
98%
Authentic
10ppm max.
7% to 10% (3 hrs) (145 to 150°C)
0.5% max. (TLC)
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
+ 9.90
165377
209.64
0.1% max.
500 g

Chemical Identifiers

5934-29-2
209.63
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
165377
C1=C(NC=N1)CC(C(=O)O)N.O.Cl
C6H12ClN3O3
MFCD00151027
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid;hydrate;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : MS3119000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only