missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Histidine, Cell Culture Reagent

$23.76 - $249.82

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-YFKPBYRVSA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin, l-histidine
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES C1=C(NC=N1)CC(C(=O)O)N
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6306509 View Documents Alfa Aesar™
J6306509
10g Each for $23.76
 
AAJ6306522 View Documents Alfa Aesar™
J6306522
100g Each for $24.99
 
AAJ6306536 View Documents Alfa Aesar™
J6306536
500g Each for $125.05
 
AAJ63065A1 View Documents Alfa Aesar™
J63065A1
1kg Each for $249.82
 
Specifications

Chemical Identifiers

71-00-1
155.157
HNDVDQJCIGZPNO-YFKPBYRVSA-N
6274
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
C6H9N3O2
MFCD00064315
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin, l-histidine
CHEBI:15971
C1=C(NC=N1)CC(C(=O)O)N

Specifications

71-00-1
MFCD00064315
14,4720
HNDVDQJCIGZPNO-YFKPBYRVSA-N
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
6274
155.16
Cell Culture Reagent
~281°C
10g
−39° (c=5 in Water)
C6H9N3O2
84088
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin, l-histidine
C1=C(NC=N1)CC(C(=O)O)N
155.157
CHEBI:15971
Powder
White
7.0 to 8
Odorless
L-Histidine
Safety and Handling

Safety and Handling

RTECSNumber : MS3070000

TSCA : TSCA