missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Glycine 7-amido-4-methylcoumarin

Chemical Identifiers

Molecular Formula C12H12N2O3
MDL Number MFCD00077122
Synonym L-Gly-AMC; L-G-AMC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65592MA View Documents Alfa Aesar™
J65592MA
10mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ65592MB View Documents Alfa Aesar™
J65592MB
25mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

C12H12N2O3
MFCD00077122
L-Gly-AMC; L-G-AMC

Specifications

Light Sensitive
10mg
MFCD00077122
Soluble in DMSO or water
Solid
White
C12H12N2O3
L-Gly-AMC; L-G-AMC
232.23
L-Glycine 7-amido-4-methylcoumarin
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C