missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Glycine 7-amido-4-methylcoumarin

Manufacturer:  Alfa Aesar™ J65592MB

 View more versions of this product

Catalog No. AAJ65592MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

L-Glycine 7-amido-4-methylcoumarin
White
C12H12N2O3
L-Gly-AMC; L-G-AMC
232.23
Light Sensitive
25mg
MFCD00077122
Soluble in DMSO or water
Solid

Chemical Identifiers

C12H12N2O3
MFCD00077122
L-Gly-AMC; L-G-AMC
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents