Please login to your online account to display your discounted pricing

L(-)-Glutathione, oxidized, hydrate, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC343840250

 View more versions of this product

Catalog No. AC343840250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Assay ≥94%
Formula Weight 612.64
Quantity 25g
Color White
Specific Rotation Condition − 99.00 (20.00°C c=4,water)
Merck Index 15, 4511
Sulfated Ash 0.1% max. (800°C)
CAS 27025-41-8
Chemical Name or Material L(-)-Glutathione, oxidized hydrate
Molecular Formula C20H32N6O12S2·xH2O
Assay Percent Range 95%
MDL Number MFCD00150701
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Packaging Glass bottle
Water 8% max. (K.F.)
Heavy Metals (as Pb) 10ppm max.
Synonym GSSG hydrate
Iron (Fe) 20ppm max.
Specific Rotation -92° to -106° (20°C, 589nm) (c=2, H2O)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material L(-)-Glutathione, oxidized hydrate
CAS 27025-41-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C20H32N6O12S2·xH2O
Formula Weight 612.64
MDL Number MFCD00150701
Synonym GSSG hydrate
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents