Please login to your online account to display your discounted pricing

γ-L-Glutamyl-p-nitroanilide, Hydrochloride, Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP26565

 View more versions of this product

Catalog No. BP26565

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 303.7
Quantity 5g
Color Beige
Molecular Formula C11H13N3O5HCl
Chemical Name or Material γ-L-Glutamyl-p-Nitroanilide, Hydrochloride
CAS 67953-08-6
ChemAlert Storage Symbol Gray
Packaging Amber Glass
Recommended Storage 0° to 5°C

Specifications

Chemical Name or Material γ-L-Glutamyl-p-Nitroanilide, Hydrochloride
CAS 67953-08-6
Molecular Formula C11H13N3O5HCl
Formula Weight 303.7
Accessories
 

0° to 5°C

SDS & Safety Documents