missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Glutamine tert-butyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H63868MD

 View more versions of this product

Catalog No. AAH63868MD


 

Description

Description

Applications
Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

Solubility
Soluble 0.3gram in 2ml water clear solution.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
Specifications

Specifications

L-Glutamine tert-butyl ester hydrochloride
98%
MFCD00039081
ZEPNUNKDKQACNC-RGMNGODLSA-N
tert-butyl (2S)-2,5-diamino-5-oxopentanoate;hydrochloride
15602445
Powder
250mg
39741-62-3
C9H19ClN2O3
H-Gln-OtBu.HCl; (S)-tert-Butyl 2,5-diamino-5-oxopentanoate hydrochloride
CC(C)(C)OC(=O)C(CCC(=O)N)N.Cl
238.712
238.71
White

Chemical Identifiers

39741-62-3
238.712
ZEPNUNKDKQACNC-RGMNGODLSA-N
15602445
C9H19ClN2O3
MFCD00039081
H-Gln-OtBu.HCl; (S)-tert-Butyl 2,5-diamino-5-oxopentanoate hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

SDS Documents