missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ L-Glutamine, 99%

$111.03 - $721.72

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6156022 View Documents Alfa Aesar™
J6156022
100g Each for $111.03
 
AAJ6156036 View Documents Alfa Aesar™
J6156036
500g Each for $407.12
 
AAJ61560A1 View Documents Alfa Aesar™
J61560A1
1kg Each for $721.72
 
Specifications

Chemical Identifiers

56-85-9
146.146
ZDXPYRJPNDTMRX-VKHMYHEASA-N
5961
(2S)-2,5-diamino-5-oxopentanoic acid
C5H10N2O3
MFCD00008044
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
CHEBI:18050
C(CC(=O)N)C(C(=O)O)N

Specifications

56-85-9
MFCD00008044
14,4471
ZDXPYRJPNDTMRX-VKHMYHEASA-N
(2S)-2,5-diamino-5-oxopentanoic acid
5961
146.14
99%
~185°C
100g
C5H10N2O3
1723797
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
C(CC(=O)N)C(C(=O)O)N
146.146
CHEBI:18050
Powder
White
5.0 to 6.0
L-Glutamine
Safety and Handling

Safety and Handling

RTECSNumber : MA2275100

TSCA : TSCA