missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

L-Glutamic Dehydrogenase, MP Biomedicals™

$241.85 - $241.85

Chemical Identifiers

CAS 9029-12-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Synonym 2r,3r,4s,5r,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, beta-d-gal, beta-d-galactopyranose, beta-d-galactose, beta-d-galactoside, beta-galactose, cerebrose, gal-beta, galactopyranose, beta-d, unii-rq6k52j67a
PubChem CID 439353
ChEBI CHEBI:27667
IUPAC Name (2R,3R,4S,5R,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)O)O)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15118980
View Documents
Mp Biomedicals
0215118980
100mg
Each for $241.85
 
Description

Description

Specifications

Chemical Identifiers

9029-12-3
180.16
2r,3r,4s,5r,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, beta-d-gal, beta-d-galactopyranose, beta-d-galactose, beta-d-galactoside, beta-galactose, cerebrose, gal-beta, galactopyranose, beta-d, unii-rq6k52j67a
CHEBI:27667
C(C1C(C(C(C(O1)O)O)O)O)O
C6H12O6
WQZGKKKJIJFFOK-UHFFFAOYNA-N
439353
(2R,3R,4S,5R,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol

Specifications

100mg
C6H12O6
WQZGKKKJIJFFOK-UHFFFAOYNA-N
(2R,3R,4S,5R,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
439353
Cloudy Suspension
9029-12-3
2r,3r,4s,5r,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, beta-d-gal, beta-d-galactopyranose, beta-d-galactose, beta-d-galactoside, beta-galactose, cerebrose, gal-beta, galactopyranose, beta-d, unii-rq6k52j67a
C(C1C(C(C(C(O1)O)O)O)O)O
180.16
CHEBI:27667
L-Glutamic Dehydrogenase
SDS
Product Certifications